• USA Home
  • C9136 - Cytochrome c from rabbit heart

EMAIL THIS PAGE TO A FRIEND
C9136 Sigma

Cytochrome c from rabbit heart

≥95% based on Mol. Wt. 12,220 basis

Purchase

Properties

Related Categories Cell Biology, Cytochrome c, Cytochromes, Electron Transport and Cellular Respiration, Metabolic Pathways,
assay   ≥95% based on Mol. Wt. 12,220 basis
storage temp.   −20°C

Description

Biochem/physiol Actions

The ready fluctuation of cytochrome c within the cell between ferrous and ferric states, makes it an efficient biological electron-transporter. It plays a vital role in cellular oxidations in both plants and animals. Generally regarded as a universal catalyst of respiration, it forms the essential electron-bridge between the respirable substrates and oxygen.

Preparation Note

Prepared with acetic acid without using TCA.

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
HA5365000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C3131 Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?