• USA Home
  • C9155 - CPSR-3 Hybri-Max from bovine plasma

EMAIL THIS PAGE TO A FRIEND
C9155 Sigma

CPSR-3 Hybri-Max from bovine plasma

sterile-filtered, hybridoma tested

Synonym: Controlled process serum replacement-type 3

Purchase

Properties

Quality Level   GMP
sterility   sterile-filtered
impurities   endotoxin, tested
suitability   hybridoma tested
shipped in   dry ice
storage temp.   −20°C

Description

Application

Designed to support the growth and cloning of myelomas and hybridomas as efficiently as FBS. Excels when used in the stabilization of newly formed hybrids in selection medium.

Preparation Note

Proprietary manufacturing process involving dialysis, adjustments to obtain an optimal protein concentration and supplementation with appropriate whole fluids of bovine origin.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?