• USA Home
  • C9224 - Anti-CD40 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
C9224 Sigma

Anti-CD40 antibody produced in goat

affinity isolated antibody, lyophilized powder

Properties

Related Categories Alphabetical Index, Analytical Cytology, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology,
species reactivity   mouse
application(s)   indirect ELISA: 0.5-1.0 μg/mL
  western blot: 0.1-0.2 μg/mL
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   mouse ... Cd40(21939)
biological source   goat
conjugate   unconjugated

Description

Immunogen

recombinant mouse CD40 extracellualr domain expressed in NSO cells.

Physical form

Lyophilized from 0.2 μm filtered solution of phosphate buffered saline

Specificity

Shows <1% cross-reactivity with recombinant human CD40.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?