• USA Home
  • C9261 - Cytochrome c from pigeon heart

EMAIL THIS PAGE TO A FRIEND
C9261 Sigma

Cytochrome c from pigeon heart

≥95% based on Mol. Wt. 12,173 basis

Purchase

Properties

assay   ≥95% based on Mol. Wt. 12,173 basis
storage temp.   −20°C

Description

Preparation Note

Prepared without using TCA.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
HA5365000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

C4011 Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?