• USA Home
  • C9267 - Cyanogen bromide-activated−Sepharose® 6MB

EMAIL THIS PAGE TO A FRIEND
C9267 Sigma

Cyanogen bromide-activated−Sepharose® 6MB

lyophilized powder, 200-300 μm

Synonym: Cyanogen bromide-activated Agarose

Purchase

Properties

Related Categories Activated/Functionalized Matrices, Affinity Chromatography, Molecular Biology, Protein Chromatography, Proteomics More...
form   lyophilized powder
matrix   Sepharose 6MB (macrobeads)
matrix active group   cyanate or related structures
matrix spacer   1 atom (when ligands are coupled via isourea derivative or related linkage)
swelling   1 g swells to ~3 mL
particle size   200-300 μm
capacity   ≥8 μmol/mL coupling capacity (6-AKS)
storage temp.   2-8°C

Description

Physical form

Lyophilized powder stabilized with lactose and dextran

Application

Cyanogen bromide-activated Sepharose is used in affinity chromatography and protein chromatography. Cyanogen bromide-activated Sepharose has been used to discover insecticidal agents, to raise polyclonal antibodies against the yeast invertase glycosyls, and to study immunological processes of xenograft rejection.

Legal Information

Sepharose is a registered trademark of GE Healthcare

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?