• USA Home
  • C9446 - Cholesteryl N-(trimethylammonioethyl)carbamate chloride

EMAIL THIS PAGE TO A FRIEND
C9446 Sigma

Cholesteryl N-(trimethylammonioethyl)carbamate chloride

~95%

Synonym: TC-Cholesterol

Purchase

Properties

Related Categories Biochemicals and Reagents, Cholesterol and Derivatives, Lipids, Sterol Lipids
assay   ~95%
shipped in   wet ice
storage temp.   −20°C

Description

Application

Cationic cholesterol derivative useful in liposome-mediated transfection.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?