• USA Home
  • C9447 - Catalase from bison liver

EMAIL THIS PAGE TO A FRIEND
C9447 Sigma

Catalase from bison liver

2,000-5,000 units/mg protein

Synonym: H2O2:H2O2 oxidoreductase

Purchase

Description

Analysis Note

Protein determined by biuret, unless otherwise indicated.

Caution

Freezing of solutions is not recommended.

Unit Definition

One unit will decompose 1.0 μmole of H2O2 per min at pH 7.0 at 25 °C, while the H2O2 conc. falls from 10.3 to 9.2 mM, measured by the rate of decrease of A240.

Price and Availability


Laboratory Gloves

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
FI4378000

Documents

Certificate of Analysis

Protocols & Articles

Protocols

Enzymatic Assay of Catalase (EC 1.11.1.6)

The continuous spectrophotometric rate reduction determination (A240, Light path = 1 cm) is based on the following reaction:
Keywords: Reductions

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

inhibitor

Product #

Description

Add to Cart

A8056 3-Amino-1,2,4-triazole, ≥95% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?