• USA Home
  • C9464 - Cholesterol Esterase from porcine pancreas

EMAIL THIS PAGE TO A FRIEND
C9464 Sigma

Cholesterol Esterase from porcine pancreas

lyophilized powder, ≥15,000 units/g protein

Synonym: Cholesterol Esterase from hog pancreas, Sterol-ester acylhydrolase

Purchase

Description

Analysis Note

Protein determined by biuret.

Other Notes

Contains potassium phosphate buffer salts and stabilizer.

Unit Definition

One unit will hydrolyze 1.0 μmole of cholesteryl oleate to cholesterol and oleic acid per min at pH 7.0 at 37 °C in the presence of taurocholate.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Cholesterol Esterification

To more efficiently transport both dietary and synthesized cholesterol, it is converted to cholesteryl esters. Free cholesterol can be taken up by lipoproteins, but is confined to the outer surface o...
BioFiles 2007, 2.7, 10.
Keywords: Biofiles, Catalysis

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?