• USA Home
  • C9618 - Monoclonal Anti-CD164 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
C9618 Sigma

Monoclonal Anti-CD164 antibody produced in mouse

~2 mg/mL, clone 67D2, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-Endolyn, Anti-MGC-24, Anti-Sialomucin CD164

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Immune Cell Signaling and Blood, Antibodies to Human Leukocytes,
species reactivity   human
application(s)   flow cytometry: 2-4 μg/test using human blood
  immunocytochemistry: suitable
  immunohistochemistry: suitable
  immunoprecipitation: suitable
  western blot: suitable
clone   67D2, monoclonal
concentration   ~2 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
mol wt   antigen mol wt 80-90 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CD164(8763)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

breast tumor cell line T-47D.

Physical form

Solution in 0.01 M phosphate buffered saline, pH7.4, containing 15 mM sodium azide.

Specificity

The antibody recognizes human CD164.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

HPA010636 Anti-CD164 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?