• USA Home
  • C9643 - Carnauba wax No. 1 yellow

EMAIL THIS PAGE TO A FRIEND
C9643 Sigma

Carnauba wax No. 1 yellow

Synonym: Brazil wax

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?