EMAIL THIS PAGE TO A FRIEND
C9652 Sigma

Carbostyril 151

Synonym: 2,7-Dihydroxy-4-trifluoromethylquinoline

  • CAS Number 73496-29-4

  • Empirical Formula (Hill Notation) C10H6F3NO2

  • Molecular Weight 229.16

  •  MDL number MFCD00057298

  •  PubChem Substance ID 24893183

Purchase

Properties

InChI Key   XAGSNZIBWDMNBS-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?