• USA Home
  • C9759 - Cardiotoxin from Naja mossambica mossambica
EMAIL THIS PAGE TO A FRIEND
C9759 Sigma
Cardiotoxin from Naja mossambica mossambica
Purchase
Description
Frequently Asked Questions
Frequently Asked Questions are available for this Product.
Other Notes
Snake toxin; mixture of cardiotoxins.
Price and Availability
Better Together
Discover your Target
Making Headway
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Certificate of Origin
Frequently Asked Questions
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis. These documents are located on the product detail page under Useful Links & Tools. Click on the following link to search for a Certificate of Analysis. Please click the following link to see the details on our Product Dating Information.
How do I get lot-specific information or a Certificate of Analysis?
A Certificate of Analysis is available by lot number and can be obtained through our Advanced Search Option: http://www.sigmaaldrich.com/catalog/AdvancedSearchPage.do
What is Product C9759, Cardiotoxin, soluble in?
Sigma tests the solubility in water at 1 mg/mL; this yields a clear to slightly hazy solution.
What is the molecular weight of Product C9759, Cardiotoxin?
According to Biochim. Biophys. Acta, 336, 470, (1974), this product is a mixture of cardiotoxins all having a molecular weight of approximately 7,000 Da.
Has this product been tested in any in vivo studies?
We have not done any bioassay studies with this product in our labs here at Sigma-Aldrich. The following references contain information regarding bioassay studies: Cell Research, 16, 713-722 (2006) and J. Thorac. Cardiovasc. Surg., 121, 871-878 (2001).
How do I find price and availability?
There are several ways to find pricing and availability for our products.  Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers. 
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS. To access the shipping information for this material, use the link on the product detail page for the product, or search here. 
My question is not addressed here, how can I contact Technical Service for assistance?
Use the option to the right to "Ask a Question" by email of a Technical Service Scientist.
Show more questions
Protocols & Articles
Peer-Reviewed Papers
15
References
Related Products
related product
Product #
Description
Add to Cart
92210 Timestrip Plus 0 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?