EMAIL THIS PAGE TO A FRIEND
C9937 Sigma

Citrazinic acid

Synonym: 2,6-Dihydroxyisonicotinic acid, 2,6-Dihydroxypyridine-4-carboxylic acid

  • CAS Number 99-11-6

  • Empirical Formula (Hill Notation) C6H5NO4

  • Molecular Weight 155.11

  •  Beilstein Registry Number 383736

  •  EC Number 202-731-2

  •  MDL number MFCD00006274

  •  PubChem Substance ID 24893207

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Building Blocks, C6, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   CSGQJHQYWJLPKY-UHFFFAOYSA-N
mp   >300 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
NS1400000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?