-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym: Acylcholine acyl-hydrolase Choline esterase, butyryl Pseudocholinesterase
CAS Number 9001-08-5
MDL number MFCD00081455
| form | lyophilized powder |
| composition | Protein, ~40% |
| storage temp. | 2-8°C |
Protein determined by biuret
One unit will hydrolyze 1.0 μmole of butyrylcholine to choline and butyrate per min at pH 8.0 at 37 °C. The activity obtained using butyrylcholine as substrate is approx. 2.5 times that obtained using acetylcholine.
Lyophilized powder containing salts
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?