• USA Home
  • CKOZFN16606 - CompoZr® Knockout ZFN Kit

EMAIL THIS PAGE TO A FRIEND
CKOZFN16606 Sigma

CompoZr® Knockout ZFN Kit

Human PEBP4 (NM_144962)

Purchase

Properties

Related Categories CompoZr Knockout ZFN Kits, Functional Genomics and RNAi, Human CompoZr Knockout ZFN Kits, Molecular Biology, PD-PLA More...
NCBI accession no.   NM_144962
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... PEBP4(157310)

Description

General description

CompoZr Knockout ZFNs create targeted double strand breaks at a gene specific target locus. Through the cellular process of Non-Homologous End Joining, this double strand break can result in modification of the DNA sequence and therefore create a functional knockout of the targeted gene. To provide the best chance of creating a functional gene knockout, CompoZr Knockout ZFNs are targeted to the first 2/3 of the coding region for the gene of interest.

Legal Information

All Zinc Finger Nucleases are sold under license from Sangamo Biosciences. For a copy of the Label License provided with purchase of CompoZr ZFNs, please visit the ZFN Label License page.

CompoZr is a registered trademark of Sigma-Aldrich Co. LLC

Physical form

CompoZr Knockout ZFNs are provided as 10 aliquots of mRNA encoding for the ZFNs along with plasmid DNA for both ZFN pairs. Also included in the kit are custom-designed primers, that allow for determination of the rate of mutation and for screening of individual clones that have the desired mutational event, and positive control DNA that has been modified using the CompoZr Knockout ZFN.

Biochem/physiol Actions

The ZFN (zinc finger nuclease) provided in this kit targets the PEBP4 (phosphatidylethanolamine binding protein 4) gene. The PEBP4 protein regulates apoptosis by associating with Raf1 (proto-oncogene serine/threonine-protein kinase) and MEK1 (MAPK/ERK kinase 1). This association blocks TNFα (tumor necrosis α) or TRAIL (tumor necrosis factor-related apoptosis-inducing ligand) - mediated activation of MEK/ERK (extracellular signal-regulated kinase). PEBP4 is also involved in myoblast differentiation. Additionally, it promotes Akt (RAC-α serine/threonine-protein kinase) activation. The gene is expressed in various cancers and is associated with cancer progression. The PEPB4 mRNA is a target of microRNA miR-34a.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

CKOZFND16606 CompoZr® Knockout ZFN Kit, ZFN plasmid only, Human PEBP4 (NM_144962)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?