• USA Home
  • CLB9 - Latex beads, carboxylate-modified polystyrene

EMAIL THIS PAGE TO A FRIEND
CLB9 Sigma

Latex beads, carboxylate-modified polystyrene

aqueous suspension, 0.9 μm mean particle size

Purchase

Properties

Related Categories Labware, Latex Beads, Latex and Glass Beads More...
form   aqueous suspension
composition   Solids, 10%
extent of labeling   ~0.1 mEq/g carboxylate per g polymer
mean particle size   0.9 μm

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?