CLS3056 Sigma

Corning® cell culture flasks

surface area 25 cm2, angled neck, cap (vented)

DOWNLOAD MSDS (PDF)

Synonym: culture flask, flask cell culture, tissue culture flask

Properties

Related Categories Cell Culture Supplies, Corning, Corning Cell Culture Flasks, Corning Flasks, Corning Plastic Cell Culture Flasks,
material   angled neck
  cap (vented)
  flask triangular
sterility   sterile
feature   graduations
packaging   case of 200
mfr. no.   Corning, 3056
maximum volume   10 mL
surface area   25 cm2
working volume   5-7.5 mL
matrix active group   TC-Treated surface treatment

Description

General description

Available in a variety of sizes, designs and cap styles to meet your needs.
• Manufactured from optically clear virgin polystyrene
• Treated for optimal cell attachment
• Printed with lot numbers for ease in traceability
• 100% integrity tested
• Sterilized by gamma irradiation and certified nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.

Price and Availability

Customers Also Viewed

Corning<SUP>®</SUP> cell culture flasks

surface area 25 cm2, angled neck, cap (phenolic-style)

Corning<SUP>®</SUP> cell culture flasks

surface area 25 cm2, angled neck, cap (phenolic-style)

Corning<SUP>®</SUP> cell culture flasks

surface area 25 cm2, canted neck, cap (plug seal)

Corning<SUP>®</SUP> cell culture flasks

surface area 25 cm2, canted neck, cap (vented)

Corning<SUP>®</SUP> cell culture flasks

surface area 75 cm2, canted neck, cap (vented)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?