CLS3059 Sigma

Corning® RoboFlask® cell culture vessels

for manual use, TC-treated, w/ bar code, Flat Cap (without septum), sterile, 50/cs

DOWNLOAD MSDS (PDF)

Synonym: cell culture chambers, cell culture flasks, cell culture vessels

Properties

Related Categories Cell Culture Supplies, Corning, Corning Flasks, Corning Plastic Cell Culture Flasks, Corning Plastic Flasks,
material   cap (flat top)
  polystyrene flask
  orange high-density polyethylene cap
description   for manual use
sterility   sterile; γ-irradiated
feature   graduations
  bar coded for efficient automated tracking
packaging   case of 50
  pack of 10
mfr. no.   Corning, 3059
maximum volume   70 mL
surface area   92.6 cm2
working volume   20-30 mL
matrix active group   TC-Treated surface treatment
suitability   suitable for (automated cell maintenance and assay systems)

Description

General description

Corning® RoboFlask® Cell Culture Vessels

Corning′s unique RoboFlask Cell Culture Vessels are designed to a standard microplate footprint (ANSI/SBS 1-2004: Microplates - Footprint Dimensions) for use in automated cell maintenance and assay systems.

Features and Benefits:

• 92.6cm2 available cell growth area
• The cap allows pipette access (up to 5 mL) or assist in harvesting of cells.
• Bar coded for efficient automated tracking
• Compatible with the Tecan Cellerity Cell Maintenance and Assay System
• Built-in vent membrane for efficient gas and liquid exchange
• Manufactured from optically clear virgin polystyrene
• Treated for optimal cell attachment
• 100% integrity tested
• Sterilized by gamma irradiation and certified nonpyrogenic
• Stackable to save valuable incubator space

Legal Information

Corning is a registered trademark of Corning, Inc.

RoboFlask is a registered trademark of Corning, Inc.

Price and Availability

Customers Also Viewed

Corning<SUP>®</SUP> RoboFlask<SUP>®</SUP> cell culture vessels

for manual use, TC-treated, w/ bar code, Flat Cap (without septum), sterile, 100/cs

Corning<SUP>®</SUP> RoboFlask<SUP>®</SUP> cell culture vessels

for automation, TC-treated, w/ bar code, Septum Cap, sterile, 100/cs

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?