• USA Home
  • CLS3827 - Corning® 384 well plate, ULA coated

EMAIL THIS PAGE TO A FRIEND
CLS3827 Sigma

Corning® 384 well plate, ULA coated

black flat bottom (clear, with lid), sterile

Purchase

Properties

material   black flat bottom (clear, with lid)
sterility   sterile
feature   lid
mfr. no.   Corning, 3827
size   384 wells
well volume   112 μL
well working volume   20-80 μL
color   black
matrix active group   Ultra-Low attachment surface treatment

Description

Legal Information

Corning is a registered trademark of Corning, Inc.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?