• USA Home
  • CLS3978XX1 - Corning® Synthemax®-R surface multiwell plates

EMAIL THIS PAGE TO A FRIEND
CLS3978XX1 Sigma

Corning® Synthemax®-R surface multiwell plates

Synthemax -R Surface 6 well plate, 9.5 cm2, polystyrene, w/ lid, individually wrapped, sterile, 2/cs, sterile

Synonym: 6 well cell culture plates, 6 well culture plates, 6 well plates, Corning cell culture plates, cell culture plates, culture plates, microplates, microtiter plates, multiwell plates

Purchase

Properties

Related Categories Cell Culture Supplies, Corning, Corning 6-Well Microplates, Corning Microplates, Culture Multiwell Plates,
material   polystyrene
  wells round
  clear wells
sterility   sterile
feature   lid
packaging   pack of 1
  case of 2
mfr. no.   Corning, 3978XX1
size   6 wells
well surface area   9.5 cm2
well volume   16.8 mL
well working volume   16.8 mL , recommended medium well volume 1-4mL
color   clear
matrix active group   surface treatment, Synthemax surface
suitability   suitable for (the differentiation of stem cells into specialized cell types)

Description

General description

Corning Synthemax cell culture surface uses an active peptide acrylate for polystyrene binding to promote stem cell proliferation and differentiation.

Corning Synthemax-R Surface is a unique synthetic surface coated onto tissue culture treated cell culture flasks and multiwell plates. The surface chemistry is designed to mimic a cells′ natural environment with an extracellular matrix derived cell adhesion promoting peptide. The peptide acrylate coating creates a uniform active surface for stem cell attachment, growth and differentiation, especially in chemically defined media.

Synthemax-R Surface multiple well plates and flasks offer a synthetic alternative to biological coatings and/or feeder cell layers used in traditional stem cell culture. The proprietary surface coating technology ensures lot-to-lot consistency, facilitating reproducible results in laboratories.

Developed by Corning scientists, this new biomimetic surface benefits are:
• Unique synthetic surface mimics natural cell environment
• Supports expansion of undifferentiated stem cells into a specialized, differentiated cell type
• Ready to use surface
• 6 well format uniformly coated
• Recommended working volume 1.9 mL to 2.9 mL
• Nonreversible lids with condensation rings to reduce contamination
• Individually wrapped
• Sterilized by gamma irradiation (SAL 10-3)
• 2 year shelf life
• Amenable to automation
• Eliminates the need and time for expensive coating

Legal Information

Corning is a registered trademark of Corning, Inc.

Synthemax is a registered trademark of Corning, Inc.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?