CLS430725 Sigma

Corning® cell culture flasks

surface area 75 cm2, canted neck, cap (phenolic-style)

DOWNLOAD MSDS (PDF)

Synonym: culture flask, flask cell culture, tissue culture flask

Properties

Related Categories Cell Culture Supplies, Corning, Corning Cell Culture Flasks, Corning Flasks, Corning Plastic Cell Culture Flasks,
material   canted neck
  cap (phenolic-style)
  flask rectangular
sterility   sterile
feature   graduations
packaging   case of 100
mfr. no.   Corning, 430725
maximum volume   60 mL
surface area   75 cm2
working volume   15-22.5 mL
matrix active group   TC-Treated surface treatment

Description

General description

Available in a variety of sizes, designs and cap styles to meet your needs.
• Manufactured from optically clear virgin polystyrene
• Treated for optimal cell attachment
• Printed with lot numbers for ease in traceability
• 100% integrity tested
• Sterilized by gamma irradiation and certified nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.

Price and Availability

Customers Also Viewed

Corning<SUP>®</SUP> cell culture flasks

surface area 25 cm2, angled neck, cap (phenolic-style)

Corning<SUP>®</SUP> cell culture flasks

surface area 25 cm2, angled neck, cap (phenolic-style)

Corning<SUP>®</SUP> cell culture flasks

surface area 75 cm2, canted neck, cap (plug seal)

Corning<SUP>®</SUP> cell culture flasks

surface area 75 cm2, canted neck, cap (vented)

Corning<SUP>®</SUP> CellBIND<SUP>®</SUP> Surface cell culture flasks

CellBIND 75cm2 rectangular canted neck cell culture flask w/ Vent Cap, PS, sterile, 100/cs

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?