• USA Home
  • CLZNS - Coelenterazine Sampler Kit

EMAIL THIS PAGE TO A FRIEND
CLZNS Sigma

Coelenterazine Sampler Kit

Purchase

Description

Components

Components:
25 μg of each of the following products C2230, C2855, C2980, C3105, C3230, C3355, C3480, C3605, C3730.

Application

These coelenterazine products differ from each other in terms of luminescence wavelength, capacity, intensity, and half-rise time. They have wide applications in calcium imaging, diagnostic immunoassays and high throughput drug screening.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?