• USA Home
  • CPS1120 - Chemiluminescent Peroxidase Substrate-1

EMAIL THIS PAGE TO A FRIEND
CPS1120 Sigma

Chemiluminescent Peroxidase Substrate-1

sufficient for 120 mL substrate

Synonym: CPS1

Purchase

Description

Application

Two component liquid peroxidase substrate system for Western blot detection. Picomole detection with superior signal-to-noise ratio. Component ratios can be adjusted for optimal signal duration, sensitivity and background.

Price and Availability


Antibody Explorer

Explore with Confidence

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?