• USA Home
  • CRD37960 - Polar Bear Plus Accessory
EMAIL THIS PAGE TO A FRIEND
CRD37960 Sigma
Polar Bear Plus Accessory
Insulated Former Kit, For 250 mL Round Bottom Flask, silicone cover
Purchase
Properties
Related Categories Cold Plates and Electronic Chillers, Cryoware, Labware More...
material   silicone cover
Description
General description
Former kit comprising high heat transfer former suitable for a 250ml round-bottom flask (not included) and an insulating cover. The former has a PTFE coating and includes a port for an external PT100 probe. We recommend our Support Rod Assembly to support the flask and/or PT100 probe. Silicone Ring, achievable temperature: -25C to 150C
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?