• USA Home
  • CRD47290 - Polar Bear Plus-Crystal, with magnetic stirring
EMAIL THIS PAGE TO A FRIEND
CRD47290 Sigma
Polar Bear Plus-Crystal, with magnetic stirring
AC input 115 V, US 2-pin plug
Purchase
Properties
Related Categories Cold Plates and Electronic Chillers, Cryoware, Labware More...
material   chrome-plated plate (copper)
measuring range   -40-150 °C temperature, accuracy: ±1.0, resolution: 0.01 °C
parameter   0, 200-1500 rpm speed
overall W × D × H   200 × 320 × 170 mm
plate diam.   149 mm
Description
Features and Benefits
• Simple operation - attach to an electricity supply and set the temperature
• Accurate and precise temperature control from -40C to +150C
• Fully programmable temperature profiles with our Polar Bear Plus Crystal
• No solvents, heat transfer fluids or ice
• Self contained and easily portable – similar in size to a shoebox
• Powerful magnetic stirring up to 1500rpm*
• Adaptable to many applications
• Large range of accessories available.
General description
Each member of the energy efficient Polar Bear Plus family accurately delivers temperatures from -40° to +150°C at the touch of a button. No solvents, ice, heat transfer fluids or other peripherals are needed – just plug it in and set the desired temperature. Sophisticated temperature control is the key to the Polar Bear Plus Crystal, developed with input from the chemists at SAFC Pharmorphix®. The temperature gradients that can be achieved, together with programmable temperature profiling and cycling, make this the first choice f! or your crystallisation investigations
Legal Information
Pharmorphix is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Polar Bear Demo [VIDEO]
Watch our video to see how easy heating and cooling is with the Polar Bear and Polar Bear Plus.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?