• USA Home
  • CS0380 - JNK 1&2 Activity Assay Kit

EMAIL THIS PAGE TO A FRIEND
CS0380 Sigma

JNK 1&2 Activity Assay Kit

sufficient for 50 reactions

Synonym: c-Jun N-terminal kinase activity assay kit

Purchase

Properties

Related Categories Cell Analysis, Cell Biology More...
usage   sufficient for 50 reactions
shipped in   dry ice
storage temp.   −20°C

Description

Application

JNK1&2 Activity Assay Kit offers an easy method to assay JNK1&2 activity and to explore new JNK stimuli, inhibitors and activators. The kit provides all the reagents required for straightforward detection and measurement of JNK activity in cell lysates, tissue homogenates, column fractions or purified kinases.The assay detects phosphorylated JNK 1&2 based on immunoprecipitation of the kinase using anti-JNK antibodies, and a detection of the phosphorylation activity of its substrate, ATF2, by immunoblotting. Alternative radioactive protocol is included.

Price and Availability


All labs need water

Kit component also available separately

Description

Product #

Add to Cart

Anti-c-Jun N-terminal Kinase .2 mL SDS    
EZviewTM Red Protein A Affinity Gel, 1 mL SDS    
ATF2 .5 mg SDS    

Kit component only

Description

Product #

Add to Cart

Monoclonal Anti-phospho ATF2 (pThr69,71)−Peroxidase 1 mL/vial SDS    
Assay Buffer For Kinase Activity 1 mL SDS    
Wash Buffer 10X 8 mL SDS    
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?