• USA Home
  • D0143 - Dansyl-ε-aminocaproic acid (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
D0143 Sigma

Dansyl-ε-aminocaproic acid (dicyclohexylammonium) salt

  • CAS Number 76563-40-1

  • Empirical Formula (Hill Notation) C18H24N2O4S · C12H23N

  • Molecular Weight 545.78

  •  PubChem Substance ID 329798587

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?