EMAIL THIS PAGE TO A FRIEND
D0163 Sigma

Dequalinium acetate

~90%

  • CAS Number 4028-98-2

  • Empirical Formula (Hill Notation) C30H40N4 · C4H6O4

  • Molecular Weight 574.75

  •  EC Number 223-712-5

Purchase

Properties

assay   ~90%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
VA0600000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

D3768 Dequalinium chloride hydrate, ≥95% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?