• USA Home
  • D0188 - Anti-Deoxyribonuclease I (DNase I) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
D0188 Sigma

Anti-Deoxyribonuclease I (DNase I) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to DNA Fragmentation,
species reactivity   bovine
application(s)   microarray: suitable
  western blot: 1 μg/mL using bovine pancreatic DNase I
clone   polyclonal
concentration   1.0 mg/mL
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt 33 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   bovine ... DNASE1(282217)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

bovine pancreatic deoxyribonuclease I (DNase I).

General description

The enzyme is Ca2+ and Mg2+ dependent. It is synthesized in the pancreas and stored in zymogen granules. It has been used to reduce the viscosity of cystic fibrosis sputum.

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 0.01% sodium azide.

Application

Anti-Deoxyribonuclease I (DNase I) antibody produced in rabbit is suitable for microarray and western blotting at a working concentration of 1 μg/mL using bovine pancreatic DNase I.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

DNase I is an endonuclease that hydrolyzes double-stranded or single-stranded DNA preferentially at sites adjacent to pyrimidine nucleotides to produce a complex mixture of 5′-phosphate mononucleotides and oligonucleotides.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?