• USA Home
  • D0268 - Dansyl-DL-α-aminocaprylic acid cyclohexylammonium salt

EMAIL THIS PAGE TO A FRIEND
D0268 Sigma

Dansyl-DL-α-aminocaprylic acid cyclohexylammonium salt

  • CAS Number 102783-22-2

  • Empirical Formula (Hill Notation) C20H28N2O4S · C6H13N

  • Molecular Weight 491.69

  •  MDL number MFCD00058044

  •  PubChem Substance ID 329798595

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?