EMAIL THIS PAGE TO A FRIEND
D0393 Sigma

Dansyl-L-citrulline

  • CAS Number 74672-25-6

  • Empirical Formula (Hill Notation) C18H24N4O5S

  • Molecular Weight 408.47

  •  MDL number MFCD00070313

  •  PubChem Substance ID 24893230

Purchase

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?