• USA Home
  • D0407 - DEAE−Cibacron Blue 3GA−Agarose

EMAIL THIS PAGE TO A FRIEND
D0407 Sigma

DEAE−Cibacron Blue 3GA−Agarose

Type 300-CL, saline suspension

Purchase

Properties

Related Categories Adsorption/Partition, Molecular Biology, Protein Chromatography, Proteomics, Silica More...
type   Type 300-CL
form   saline suspension
extent of labeling   0.2-0.4 μmol per mL
matrix   cross-linked agarose
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Price and Availability


Amplified Detection

Amicon Pro Purification System
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?