• USA Home
  • D043 - (S)-(+)-Apomorphine hydrochloride hydrate

EMAIL THIS PAGE TO A FRIEND
D043 Sigma

(S)-(+)-Apomorphine hydrochloride hydrate

solid

Synonym: S(+)-APO hydrochloride, S(+)-10,11-Dihydroxyaporphine hydrochloride

Purchase

Properties

Related Categories Antagonists, Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Dopaminargics - Antagonists,
InChI Key   SKYZYDSNJIOXRL-ZOWNYOTGSA-N
assay   ≥98% (HPLC)
form   solid
optical activity   [α]23/D +74.5°, c = 0.15 in methanol(lit.)
storage condition   desiccated
  protect from light
  under inert gas
color   white to light gray
solubility   H2O: ≥10 mg/mL
  ethanol: soluble20 mg/mL
storage temp.   2-8°C

Description

Biochem/physiol Actions

Dopamine receptor antagonist; inactive as an agonist.

Caution

Hygroscopic, photosensitive, may decompose and turn green on exposure to light and air.
Reseal under argon or nitrogen to maximize stability.

Price and Availability


Neuroscience Grant Programm
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?