• USA Home
  • D045 - 1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine hydrochloride

EMAIL THIS PAGE TO A FRIEND
D045 Sigma

1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine hydrochloride

Synonym: MPTP hydrochloride

  • CAS Number 23007-85-4

  • Empirical Formula (Hill Notation) C12H15N · HCl

  • Molecular Weight 209.72

  •  Beilstein Registry Number 3707312

  •  MDL number MFCD00012790

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   KOWJANGMTAZWDT-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Dopaminergic neurotoxin.

Warning

Reportedly causes a severe and irreversible Parkinsonian condition in humans and monkeys.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3
RTECS 
UT8358950

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

M0896 1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine hydrochloride, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?