• USA Home
  • D0510 - 2,2′-Dithiodibenzoic acid

EMAIL THIS PAGE TO A FRIEND
D0510 Sigma

2,2′-Dithiodibenzoic acid

~95%

Synonym: 2,2′-Di(thiosalicylic acid), Bis(2-carboxyphenyl) disulfide

Purchase

Properties

assay   ~95%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
DG9660000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?