• USA Home
  • D0536 - Anti-Donkey Serum antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
D0536 Sigma

Anti-Donkey Serum antibody produced in rabbit

whole antiserum

Synonym: Anti-Serum, donkey

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Animal Plasma Proteins, DM-DP, Primary Antibodies,
biological source   rabbit
antibody form   whole antiserum
clone   polyclonal
contains   15 mM sodium azide
species reactivity   donkey
application(s)   immunoelectrophoresis: suitable
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

General description

Pooled normal donkey serum was used as an immunogen in rabbit to develop anti-donkey whole serum antibody.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?