• USA Home
  • D0635 - N6,2′-O-Disuccinyladenosine 3′:5′-cyclic monophosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
D0635 Sigma

N6,2′-O-Disuccinyladenosine 3′:5′-cyclic monophosphate sodium salt

~98%

Synonym: Disuccinyl-cAMP

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Type, Enzymes, Inhibitors, and Substrates, Substrate Analogs More...
InChI Key   ZIKFESRAKKTODM-UHFFFAOYSA-N
assay   ~98%
storage temp.   −20°C

Description

Application

Starting material for synthesis of a wide range of cyclic-AMP derivatives: chemiluminescent conjugates, affinity matrices, etc.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?