• USA Home
  • D0648 - 5′-O-(4,4′-Dimethoxytrityl)thymidine 3′-(2-chlorophenyl) phosphate triethylammonium salt

EMAIL THIS PAGE TO A FRIEND
D0648 Sigma

5′-O-(4,4′-Dimethoxytrityl)thymidine 3′-(2-chlorophenyl) phosphate triethylammonium salt

~98%

Purchase

Properties

assay   ~98%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?