• USA Home
  • D0696 - DCAMKL2 (1-690), active, GST tagged human

EMAIL THIS PAGE TO A FRIEND
D0696 Sigma

DCAMKL2 (1-690), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: DCDC3, DCLK2, DKFZp761I032, MGC45428

Purchase

Properties

Related Categories Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in baculovirus infected Sf9 cells
grade   PRECISIO® Kinase
assay   ≥70% (SDS-PAGE)
form   buffered aqueous glycerol solution
activity   80-108 nmol/min·mg
mol wt   mol wt ~105 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... DCLK2(166614)

Description

Biochem/physiol Actions

DCAMKL2 (also known as doublecortin-like kinase 2 or DCLK2) is a close homologue of doublecortin or DCX which is associated with intractable epilepsy in humans, due to a severe disorganization of the neocortex and hippocampus known as classical lissencephaly. DCX; DCLK2-null mice display frequent spontaneous seizures that originate in the hippocampus, with most animals dying in the first few months of life. DCX and DCLK2 are co-expressed in developing hippocampus, and, in their absence, there is dosage-dependent disrupted hippocampal lamination associated with a cell-autonomous simplification of pyramidal dendritic arborizations leading to reduced inhibitory synaptic tone.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?