• USA Home
  • D0763 - N,N-Dimethyl-p-nitrosoaniline

EMAIL THIS PAGE TO A FRIEND
D0763 Sigma

N,N-Dimethyl-p-nitrosoaniline

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1369 4.2 / PGII
WGK Germany 
3
RTECS 
BX7175000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

D172405 N,N-Dimethyl-4-nitrosoaniline, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?