• USA Home
  • D0884 - Dnase/collagenase mix

EMAIL THIS PAGE TO A FRIEND
D0884 Sigma

Dnase/collagenase mix

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?