• USA Home
  • D1028 - P1,P4-Di(guanosine-5′) tetraphosphate ammonium salt

EMAIL THIS PAGE TO A FRIEND
D1028 Sigma

P1,P4-Di(guanosine-5′) tetraphosphate ammonium salt

~95% (HPLC)

Synonym: Diguanosine tetraphosphate, G(5′)P4(5′)G, GP4G

  • CAS Number 102783-33-5

  • Empirical Formula (Hill Notation) C20H28N10O21P4

  • Molecular Weight 868.39

  •  MDL number MFCD00058645

  •  PubChem Substance ID 24893328

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotide Analogs More...
InChI Key   WDOHMQPAFMGSPZ-UHFFFAOYSA-O
assay   ~95% (HPLC)
storage temp.   −20°C

Description

Packaging

1 mg in poly bottle

Application

P1,P4-Di(guanosine-5′) tetraphosphate (Diguanosine tetraphosphate) (GP4G) is a dinucleoside polyphosphate which may be used to study potential pathological effects of dinucleoside polyphosphtes in vivo. GP4G is an acceptor molecule for cytidine-3′, 5′-bisphosphate from the adenylylated donor 5′-adenylylated cytidine 5′,3′-bisphosphate in the T4 RNA ligase catalyzed reactions. GP4G is a used as an inhibitor of DNA apurinic/apyrimidinic endonuclease (APE1).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?