• USA Home
  • D1036 - 4,5-Diamino-2-hydroxy-6-mercaptopyrimidine hemisulfate salt

EMAIL THIS PAGE TO A FRIEND
D1036 Sigma

4,5-Diamino-2-hydroxy-6-mercaptopyrimidine hemisulfate salt

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Type, Enzymes, Inhibitors, and Substrates, Substrate Analogs More...
InChI Key   CPMIQTKEFQJCPM-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?