• USA Home
  • D1037 - Dextran from Leuconostoc mesenteroides

EMAIL THIS PAGE TO A FRIEND
D1037 Sigma

Dextran from Leuconostoc mesenteroides

average mol wt 425,000-575,000

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Dextrans, Dextrans for General Research Applications
Quality Level   PREMIUM
mol wt   average mol wt 425,000-575,000

Description

Application

Use of dextrans as long and hydrophilic spacer arms improves the performance of immobilized proteins acting on macromolecules.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Articles

Dextran and Related Polysaccharides

Historically, dextrans had been long recognized as contaminants in sugar processing and other food production. The formation of dextran in wine was shown by Pasteur to be due to the activity of micro...
Vicki Caligur
BioFiles 2008, 3.10, 17.
Keywords: Adhesion, Applications, Chromatography, Clinical, Cosmetics, Derivatizations, Diffusion, Filtration, Immobilization, Industries, Ion Exchange, Ligands, Microscopy, Molecular biology, Molecular biology techniques, Nuclear magnetic resonance spectroscopy, Nucleic acid hybridization, Pharmaceutical, Precipitation, Purification, Separation, Size-exclusion chromatography, Spectroscopy, Sulfations, Type

Protocols

Dextran

Dextran is a polymer of anhydroglucose.  It is composed of approximately 95% alpha-D-(1-6) linkages. The remaining a(1-3) linkages account for the branching of dextran.1,2,3 Conflicting data on the b...
Keywords: Centrifugation, Diffusion, Fractionation, Immunoelectrophoresis, Precipitation, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?