• USA Home
  • D1165 - Dihydroxyeicosatetraenoic acid mixture for HPLC

EMAIL THIS PAGE TO A FRIEND
D1165 Sigma

Dihydroxyeicosatetraenoic acid mixture for HPLC

100 μg/mL in ethanol, >95% (each component)

Purchase

Properties

assay   >95% (each component)
concentration   100 μg/mL in ethanol
shipped in   dry ice
storage temp.   −20°C

Description

Other Notes

Approx. equal amounts of Leukotriene B4, trans-6-Leukotriene B4, trans-6-12-epi-Leukotriene B4, (5S,6R)-Di-HETE, and (5S,6S)-Di-HETE

Packaging

Packaged under Argon.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D9412 (5S,6S)-Dihydroxy-(7E,9E,11Z,14Z)-eicosatetraenoic acid, ≥90%, ~100 μg/mL in ethanol

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?