EMAIL THIS PAGE TO A FRIEND
D1630 Sigma

DNP-sarcosine

Synonym: N-(2,4-Dinitrophenyl)-sarcosine

  • CAS Number 3129-54-2

  • Empirical Formula (Hill Notation) C9H9N3O6

  • Molecular Weight 255.18

  •  MDL number MFCD00057600

  •  PubChem Substance ID 24893500

Purchase

Properties

Related Categories Amino Acid Analysis, Amino Acid Analysis Reagents, Biochemicals and Reagents, Peptide Analysis and Characterization, Peptides More...
InChI Key   SDUKWLYCRWBFJD-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?