• USA Home
  • D1759 - 5-Diazo-2′-deoxyuridine

EMAIL THIS PAGE TO A FRIEND
D1759 Sigma

5-Diazo-2′-deoxyuridine

~95%

Purchase

Properties

assay   ~95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?