• USA Home
  • D1762 - P1,P5-Di(guanosine-5′) pentaphosphate ammonium salt

EMAIL THIS PAGE TO A FRIEND
D1762 Sigma

P1,P5-Di(guanosine-5′) pentaphosphate ammonium salt

Synonym: Diguanosine pentaphosphate, G(5′]P5(5′)G

  • CAS Number 102783-42-6

  • Empirical Formula (Hill Notation) C20H29N10O24P5

  • Molecular Weight 948.37

  •  MDL number MFCD11045927

  •  PubChem Substance ID 24893537

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Type, Enzymes, Inhibitors, and Substrates, Substrate Analogs More...
InChI Key   JYGFCNUGRSNROE-AORRWANRSA-N
storage temp.   −20°C

Description

Biochem/physiol Actions

Naturally occurring dinucleoside polyphosphate, approx. 10 nM in normal human plasma. At physiological concentrations, potentiates angiotensin-induced vasoconstriction, possibly via activation of rho kinase through P2Y receptors, and increases blood pressure.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?