• USA Home
  • D1776 - N,N-Dimethylglycine ethyl ester

EMAIL THIS PAGE TO A FRIEND
D1776 Sigma

N,N-Dimethylglycine ethyl ester

Purchase

Properties

density   0.93 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
RTECS 
MB9990000
Flash Point(F) 
89.6 °F
Flash Point(C) 
32 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?