• USA Home
  • D1918 - (S)-Diacetyl Reductase from baker's yeast (S. cerevisiae)

EMAIL THIS PAGE TO A FRIEND
D1918 Sigma

(S)-Diacetyl Reductase from baker's yeast (S. cerevisiae)

lyophilized powder, 0.25-0.5 unit/mg solid

Purchase

Properties

form   lyophilized powder
storage temp.   2-8°C

Description

Biochem/physiol Actions

Enantioselectively and regioselectively reduces diketones, oxoaldehydes, methyl ketones, oxoesters and cyclohexanediones to their S-configuration hydroxy-derivatives.

Unit Definition

One unit will convert 1 μmole of diacetyl and NADPH to (S)-acetoin and NADP per min at pH 6.9 at 25°C. Enantioselectively and regioselectively reduces diketones, oxoaldehydes, methyl ketones, oxoesters and cyclohexanediones to their S-configuration

Physical form

Lyophilized powder containing Tris and DTT

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?